Mouse Recombinant CD40L/CD154/TNFSF5 Protein (Fc Tag) (from HEK293 Cells)
Supplier: Sino Biological
50327-M01H-50
103667-254EA 199.98 USD
103667-254
Mouse Recombinant CD40L/CD154/TNFSF5 Protein (Fc Tag) (from HEK293 Cells)
Proteins and Peptides
A DNA sequence encoding the soluble form of mouse CD40LG (P27548) (Gly 115-Leu 260) was fused with the Fc region of human IgG1 at the N-terminus.