Mouse Recombinant SerpinF1/PEDF Protein (His Tag) (from HEK293 Cells)
Supplier: Sino Biological
50235-M08H-20 50235-M08H-50
103667-152EA 148.35 USD
103667-152 103667-154
Mouse Recombinant SerpinF1/PEDF Protein (His Tag) (from HEK293 Cells)
Proteins and Peptides
A DNA sequence encoding the extracellular domain (Met 1-Thr 417) of mouse SerpinF1 (NP_035470.3) precursor was expressed with a C-terminal polyhistidine tag.