Zika virus (ZIKV) (strain Zika SPH2016) ZIKV-E (Stem/anchor domain of flavivirus envelope glycoprotein E) Protein (Fc Tag) (from HEK293 Cells)
Supplier: Sino Biological
40543-V02H3-100
103633-518EA 351.48 USD
103633-518
Zika virus (ZIKV) (strain Zika SPH2016) ZIKV-E (Stem/anchor domain of flavivirus envelope glycoprotein E) Protein (Fc Tag) (from HEK293 Cells)
Proteins and Peptides
A DNA sequence encoding the Zika virus (strain Zika SPH2016) E_stem (Stem/anchor domain of flavivirus envelope glycoprotein E) (ALU33341.1) (Gly698-Ala794) was expressed with the Fc region of human IgG1 at the C-terminus.