Influenza B (B/PHUKET/3073/2013) Recombinant Hemagglutinin/HA1 Protein (His Tag) (from HEK293 Cells)
Supplier: Sino Biological
40498-V08H1-100
103633-750EA 351.48 USD
103633-750
Influenza B (B/PHUKET/3073/2013) Recombinant Hemagglutinin/HA1 Protein (His Tag) (from HEK293 Cells)
Proteins and Peptides
A DNA sequence encoding the Influenza B virus ((B/PHUKET/3073/2013) hemagglutinin (Translated amino acids of EPI529345) (Met1-Arg361), termed as HA1, was expressed with a polyhistidine tag at the C-terminus.