HIV-1 (group M, subtype A, isolate 92RW020) Recombinant Envelope glycoprotein gp160 (gp120 subunit) Protein (His Tag) (from HEK293 Cells)
Supplier: Sino Biological
40403-V08H-100
103663-648EA 351.48 USD
103663-648
HIV-1 (group M, subtype A, isolate 92RW020) Recombinant Envelope glycoprotein gp160 (gp120 subunit) Protein (His Tag) (from HEK293 Cells)
Proteins and Peptides
A DNA sequence encoding the HIV-1 envelope glycoprotein gp160 (AAT67478.1) extracellular domain (Glu31-Arg494),termed as gp120, was expressed with a polyhistidine tag at the C-terminus.