Influenza A H4N2 (A/duck/Hunan/8-19/2009) Recombinant Hemagglutinin/HA Protein (His Tag) (from Baculovirus Insect Cells)
Supplier: Sino Biological
40390-V08B-100
103664-094EA 351.48 USD
103664-094
Influenza A H4N2 (A/duck/Hunan/8-19/2009) Recombinant Hemagglutinin/HA Protein (His Tag) (from Baculovirus Insect Cells)
Proteins and Peptides
A DNA sequence encoding the Influenza A virus (A/duck/Hunan/8-19/2009(H4N2)) hemagglutinin (ADN78261.1) (Met1-Asp527), termed as HA, was expressed with a polyhistidine tag at the C-terminus