Rift Valley fever virus (RVFV) (strain MP12) Recombinant glycoprotein/G1 Protein (His Tag) (from Baculovirus Insect Cells)
Supplier: Sino Biological
40338-V08B-100 40338-V08B-100
103906-158EA 351.48 USD
CA76479-818 103906-158
Rift Valley fever virus (RVFV) (strain MP12) Recombinant glycoprotein/G1 Protein (His Tag) (from Baculovirus Insect Cells)
Proteins and Peptides
A DNA sequence encoding the Rift Valley fever virus (RVFV) (strain MP12) GP (ABD38821.1) (Pro156-Thr581) was fused with a polyhistidine tag at the C-terminus.