Human Recombinant immunodeficiency virus 1 (HIV-1) (group M, subtype C, strain 92BR025) gp140 Protein (Fc Tag) (from HEK293 Cells)
Supplier: Sino Biological
40251-V02H-100
103663-976EA 351.48 USD
103663-976
Human Recombinant immunodeficiency virus 1 (HIV-1) (group M, subtype C, strain 92BR025) gp140 Protein (Fc Tag) (from HEK293 Cells)
Proteins and Peptides
A DNA sequence encoding the Human immunodeficiency virus 1 (HIV1) (group M, subtype C, strain 92BR025) GP140 (AAB61124.1) (Asn32-Ile677) was expressed with the Fc region of human IgG1 at the C-terminus.