Influenza A H7N2 (A/ruddy turnstone/New Jersey/563/2006) Recombinant Hemagglutinin/HA Protein (His Tag) (from Baculovirus Insect Cells)
Supplier: Sino Biological
40170-V08B-100
103663-890EA 351.48 USD
103663-890
Influenza A H7N2 (A/ruddy turnstone/New Jersey/563/2006) Recombinant Hemagglutinin/HA Protein (His Tag) (from Baculovirus Insect Cells)
Proteins and Peptides
A DNA sequence encoding the Influenza A virus (A/ruddy turnstone/New Jersey/563/2006(H7N2)) hemagglutinin (ACS68445.1) (Met1-Asp523) was expressed with a C-terminal polyhistidine tag.