Influenza A H1N9 (A/mallard/Ohio/265/1987) Recombinant Hemagglutinin/HA Protein (His Tag) (from Baculovirus Insect Cells)
Supplier: Sino Biological
40136-V08B-100
103663-796EA 351.48 USD
103663-796
Influenza A H1N9 (A/mallard/Ohio/265/1987) Recombinant Hemagglutinin/HA Protein (His Tag) (from Baculovirus Insect Cells)
Proteins and Peptides
A DNA sequence encoding the Influenza A virus(A/mallard/Ohio/265/1987 (H1N9)) hemagglutinin (ABK40634.1)(Met1-Gln529), termed as HA, was expressed with a polyhistidine tag at the C-terminus.