Influenza A H7N9 (A/Anhui/1/2013) Recombinant Matrix 1/M1 Protein (His Tag) (from E. coli)
Supplier: Sino Biological
40107-V08E-100
103663-744EA 401.98 USD
103663-744
Influenza A H7N9 (A/Anhui/1/2013) Recombinant Matrix 1/M1 Protein (His Tag) (from E. coli)
Proteins and Peptides
A DNA sequence encoding the Influenza A virus (A/Anhui/1/2013(H7N9)) Matrix protein 1 (EPI439506) (Met1-Lys252) was expressed with a C-terminal polyhistidine tag.