Influenza A H12N1 (A/mallard duck/Alberta/342/1983) Recombinant Hemagglutinin/HA Protein (His Tag) (from HEK293 Cells)
Supplier: Sino Biological
40029-V08H-100
103663-542EA 351.48 USD
103663-542
Influenza A H12N1 (A/mallard duck/Alberta/342/1983) Recombinant Hemagglutinin/HA Protein (His Tag) (from HEK293 Cells)
Proteins and Peptides
A DNA sequence encoding the extracellular domain of influenza A virus hemagglutinin (A/mallard duck/Alberta/342/1983 (H12N1)) (ABB88099.1) (Met 1-Lys 527) was fused with a polyhistidine tag at the C-terminus.