Human Recombinant B7H7/HHLA2 Protein (ECD, Fc Tag) (from HEK293 Cells)
Supplier: Sino Biological
16139-H02H-50 16139-H02H-200
103663-394EA 481.6 USD
103663-394 103614-956
Human Recombinant B7H7/HHLA2 Protein (ECD, Fc Tag) (from HEK293 Cells)
Proteins and Peptides
A DNA sequence encoding the human HHLA2 (NP_009003.1) (Met1-Asn344) was expressed with the Fc region of human IgG1 at the C-terminus.