Human Recombinant LILRB1/CD85j/ILT2 Protein (ECD, Fc Tag) (from HEK293 Cells)
Supplier: Sino Biological
16014-H02H-50 16014-H02H-200
103614-832EA 179.78 USD
103614-832 103663-288
Human Recombinant LILRB1/CD85j/ILT2 Protein (ECD, Fc Tag) (from HEK293 Cells)
Proteins and Peptides
A DNA sequence encoding the human LILRB1 (ADJ55949.1) (Met1-His458) was expressed with the Fc region of human IgG1 at the C-terminus.