Human/Cynomolgus Recombinant FGF16/FGF-16 Protein (from Baculovirus Insect cells)
Supplier: Sino Biological
16010-HNAB-100 16010-HNAB-5 16010-HNAB-1 16010-HNAB-1 16010-HNAB-20
103614-820EA 139.38 USD
103614-820 103677-728 103663-274 103677-730 CA76453-748
Human/Cynomolgus Recombinant FGF16/FGF-16 Protein (from Baculovirus Insect cells)
Proteins and Peptides
A DNA sequence encoding the human / cynomolgus FGF16 [(Identical to the huamn FGF16 (NP_003859.1)] (Met1-Arg207) was expressed. Human and Cynomolgus FGF16 sequences are identical.