Human Recombinant LRRTM2 (His Tag) (from HEK293 Cells)
Supplier: Sino Biological
15698-H08H-100
103663-228EA 351.48 USD
103663-228
Human Recombinant LRRTM2 (His Tag) (from HEK293 Cells)
Proteins and Peptides
A DNA sequence encoding the human LRRTM2 (NP_056379.1) (Met1-Arg422) was expressed with a polyhistidine tag at the C-terminus.