Human Recombinant OBP2B/Odorant-binding protein 2b Protein (His Tag) (from HEK293 Cells)
Supplier: Sino Biological
15608-H08H-20 15608-H08H-100
103614-600EA 148.35 USD
103614-600 103663-164
Human Recombinant OBP2B/Odorant-binding protein 2b Protein (His Tag) (from HEK293 Cells)
Proteins and Peptides
A DNA sequence encoding the human OBP2B (NP_055396.1) (Met1-His170) was expressed with a polyhistidine tag at the C-terminus.