Human Recombinant LAP3/Cytosol aminopeptidase Protein (His Tag) (from HEK293 Cells)
Supplier: Sino Biological
15480-H08H-20 15480-H08H-100
103614-690EA 148.35 USD
103614-690 103663-058
Human Recombinant LAP3/Cytosol aminopeptidase Protein (His Tag) (from HEK293 Cells)
Proteins and Peptides
A DNA sequence encoding the human LAP3 (NP_056991.2) (Met1-Ala519) was expressed with a polyhistidine tag at the C-terminus.