Human Recombinant PLS3/Plastin 3 Protein (His Tag) (from E. coli)
Supplier: Sino Biological
15042-H07E-100 15042-H07E-20
103614-268EA 148.35 USD
103614-268 103662-884
Human Recombinant PLS3/Plastin 3 Protein (His Tag) (from E. coli)
Proteins and Peptides
A DNA sequence encoding the human PLS3 (P13797) (Gly102-Asn375) was expressed with a polyhistide tag at the N-terminus.