Human Recombinant GCAP1/GUCA1A Protein (from Baculovirus Insect cells)
Supplier: Sino Biological
14565-HNCB-20 14565-HNCB-100
103662-744EA 351.48 USD
103662-744 103616-428
Human Recombinant GCAP1/GUCA1A Protein (from Baculovirus Insect cells)
Proteins and Peptides
A DNA sequence encoding the human GUCA1A (P43080)(Met1-Gly201) was expressed and purified with two additional amino acids (Gly and Pro) at the N-terminus.