Human Recombinant SLITRK6 Protein (His Tag) (from HEK293 Cells)
Supplier: Sino Biological
13922-H08H-20 13922-H08H-50
103614-312EA 300.98 USD
103614-312 103614-310
Human Recombinant SLITRK6 Protein (His Tag) (from HEK293 Cells)
Proteins and Peptides
A DNA sequence encoding the human SLITRK6 (Q9H5Y7) (Met1-Ser608) was expressed with a polyhistidine tag at the C-terminus.