Human Recombinant GPHA2/Glycoprotein hormone alpha 2 Protein (His Tag) (from HEK293 Cells)
Supplier: Sino Biological
13894-H08H-100 13894-H08H-20
103624-482EA 351.48 USD
103624-482 103614-250
Human Recombinant GPHA2/Glycoprotein hormone alpha 2 Protein (His Tag) (from HEK293 Cells)
Proteins and Peptides
A DNA sequence encoding the human GPHA2 (Q96T91) (Met1-Tyr129) was expressed with a polyhistidine tag at the C-terminus.