Human Recombinant IL23R/IL23 Receptor Protein (His Tag) (from HEK293 Cells)
Supplier: Sino Biological
13840-H08H-100 13840-H08H-100
103905-990EA 300.98 USD
103905-990 CA76479-650
Human Recombinant IL23R/IL23 Receptor Protein (His Tag) (from HEK293 Cells)
Proteins and Peptides
A DNA sequence encoding the human IL23R (AAM44229.1) (Met1-Asp353) was expressed with a polyhistidine tag at the C-terminus.