Human Recombinant LAMP2/CD107b Protein (His Tag) (from HEK293 Cells)
Supplier: Sino Biological
13555-H08H-200 13555-H08H-50
103624-306EA 481.6 USD
103624-306 103614-192
Human Recombinant LAMP2/CD107b Protein (His Tag) (from HEK293 Cells)
Proteins and Peptides
A DNA sequence encoding the human LAMP2 (NP_054701.1) (Met1-Ile375) was expressed with a C-terminal polyhistidine tag.