Human Recombinant ZG16/ZG16P Protein (Fc Tag) (from HEK293 Cells)
Supplier: Sino Biological
13450-H05H-20 13450-H05H-100
103616-364EA 148.35 USD
103616-364 103623-370
Human Recombinant ZG16/ZG16P Protein (Fc Tag) (from HEK293 Cells)
Proteins and Peptides
A DNA sequence encoding the human ZG16 (O60844.1) (Met1-Cys167) was expressed with the Fc region of mouse IgG1 at the C-terminus.