Human Recombinant NGFR/P75 Protein (Fc Tag) (from HEK293 Cells)
Supplier: Sino Biological
13184-H02H-50 13184-H02H-100
103616-322EA 191.35 USD
103616-322 103623-290
Human Recombinant NGFR/P75 Protein (Fc Tag) (from HEK293 Cells)
Proteins and Peptides
A DNA sequence encoding the human NGFR (P08138) extracellular domain (Met 1-Asn 250) was fused with the Fc region of human IgG1 at the C-terminus.