Human Recombinant CCDC134 (His Tag) (from HEK293 Cells)
Supplier: Sino Biological
13168-H08H-100
103623-276EA 351.48 USD
103623-276
Human Recombinant CCDC134 (His Tag) (from HEK293 Cells)
Proteins and Peptides
A DNA sequence encoding the human CCDC134 (Q9H6E4) (Met 1-Leu 229) was fused with a polyhistidine tag at the C-terminus.