Human Recombinant CGB7 Protein (His Tag) (from Baculovirus Insect cells)
Supplier: Sino Biological
13120-H08B-100 13120-H08B-20
103623-258EA 351.48 USD
103623-258 103613-216
Human Recombinant CGB7 Protein (His Tag) (from Baculovirus Insect cells)
Proteins and Peptides
A DNA sequence encoding the human CGB7 isoform 1 (P01233-1) (Met 1-Gln 165) was fused with a polyhistidine tag at the C-terminus.