Human Recombinant SELP/selectin P/P-selectin Protein (ECD, His Tag) (from HEK293 Cells)
Supplier: Sino Biological
13025-H08H-100
103623-068EA 250.48 USD
103623-068
Human Recombinant SELP/selectin P/P-selectin Protein (ECD, His Tag) (from HEK293 Cells)
Proteins and Peptides
A DNA sequence encoding the human SELP (AAN06828.1) extracellular domain (Met 1-Ala 771) was fused with a polyhistidine tag at the C-terminus.