Human Recombinant MAX/MYC associated factor X Protein (His & GST Tag) (from Baculovirus Insect cells)
Supplier: Sino Biological
12885-H20B-20 12885-H20B-50
103616-110EA 116.1 USD
103616-110 103616-112
Human Recombinant MAX/MYC associated factor X Protein (His & GST Tag) (from Baculovirus Insect cells)
Proteins and Peptides
A DNA sequence encoding the human MAX (NP_002373) (Met1-Ser160) was fused with the N-terminal polyhistidine-tagged GST tag at the N-terminus.