Human Recombinant Mucin-1/MUC-1 Protein (Fc Tag) (from HEK293 Cells)
Supplier: Sino Biological
12123-H02H-50 12123-H02H-100
103622-978EA 300.98 USD
103622-978 103613-024
Human Recombinant Mucin-1/MUC-1 Protein (Fc Tag) (from HEK293 Cells)
Proteins and Peptides
A DNA sequence encoding the human MUC1 isoform 2 (NP_001018016.1) extracellular domain (Met 1-Gly 167) was fused with the Fc region of human IgG1 at the C-terminus.