Human Recombinant Pepsinogen C/PGC Protein (His Tag) (from E. coli)
Supplier: Sino Biological
12072-H07E-50 12072-H07E-20
103612-994EA 148.35 USD
103612-994 103612-996
Human Recombinant Pepsinogen C/PGC Protein (His Tag) (from E. coli)
Proteins and Peptides
A DNA sequence encoding the mature form of human PGC (P20142-1) (Ile153-Ile239) was expressed with a polyhistide tag at the N-terminus.