Human Recombinant CAMK1/CaMKI-alpha Protein (from E. coli)
Supplier: Sino Biological
11932-HNCE-50 11932-HNCE-20
103612-880EA 199.98 USD
103612-880 103612-878
Human Recombinant CAMK1/CaMKI-alpha Protein (from E. coli)
Proteins and Peptides
A DNA sequence encoding the human CAMK1 (NP_003647.1) (Leu 2-Leu 370) was expressed and purified, with two additional amino acids (Gly and Pro) at the N-terminus.