Human Recombinant Neurexin-3-beta/NRXN3 Protein (His Tag) (from HEK293 Cells)
Supplier: Sino Biological
11843-H08H-50 11843-H08H-100
103612-814EA 191.35 USD
103612-814 103622-822
Human Recombinant Neurexin-3-beta/NRXN3 Protein (His Tag) (from HEK293 Cells)
Proteins and Peptides
A DNA sequence encoding the human NRXN3 beta isoform 2 (NP_620426.2) extracellular domain (Met 1-Thr 357) was expressed, with a polyhistidine tag at the C-terminus.