Human Recombinant CD300C/CMRF-35/IGSF16 Protein (His Tag) (from HEK293 Cells)
Supplier: Sino Biological
11832-H08H-100 11832-H08H-50
103622-816EA 300.98 USD
103622-816 103613-524
Human Recombinant CD300C/CMRF-35/IGSF16 Protein (His Tag) (from HEK293 Cells)
Proteins and Peptides
A DNA sequence encoding the human CD300C (NP_006669.1) extracellular domain (Met 1-Arg 183) was expressed, with a polyhistidine tag at the C-terminus.