Influenza A H4N6 (A/mallard/Ohio/657/2002) Recombinant Hemagglutinin Protein (HA1 Subunit) (His Tag) (from HEK293 Cells)
Supplier: Sino Biological
11714-V08H1-100
103632-736EA 351.48 USD
103632-736
Influenza A H4N6 (A/mallard/Ohio/657/2002) Recombinant Hemagglutinin Protein (HA1 Subunit) (His Tag) (from HEK293 Cells)
Proteins and Peptides
A DNA sequence encoding the N-terminal segment (Met 1-Arg 343) of the influenza A H4N6 hemagglutinin (A/mallard/Ohio/657/2002 (H4N6)) (ABI47995.1), termed as HA1, was fused with a C-terminal polyhistidine tag.