Influenza A H3N2 (A/Aichi/2/1968) Recombinant Hemagglutinin/HA (ECD, His Tag) (from HEK293 Cells)
Supplier: Sino Biological
11707-V08H-100
103622-664EA 351.48 USD
103622-664
Influenza A H3N2 (A/Aichi/2/1968) Recombinant Hemagglutinin/HA (ECD, His Tag) (from HEK293 Cells)
Proteins and Peptides
A DNA sequence encoding the extracellular domain (Met 1-Trp 530) of the influenza A hemagglutinin (A/Aichi/2/1968 (H3N2)) (AAA43178.1) (HA1+HA2, uncleaved) was expressed, fused with a C-terminal polyhistidine tag.