Human Recombinant SULT1A1/STP1 Protein (His Tag) (from E. coli)
Supplier: Sino Biological
11658-H07E-50 11658-H07E-20
103612-518EA 191.35 USD
103612-518 103612-520
Human Recombinant SULT1A1/STP1 Protein (His Tag) (from E. coli)
Proteins and Peptides
A DNA sequence encoding the human SULT1A1 (P50225-1) (Glu 2-Leu 295) was fused with a polyhistidine tag at the N-terminus.