Human Recombinant ARG1/Arginase 1 Protein (His & MYC Tag) (from HEK293 Cells)
Supplier: Sino Biological
11558-H35H-50 11558-H35H-50 11558-H35H-10
103681-304EA 351.48 USD
103681-304 103681-302 CA76456-754
Human Recombinant ARG1/Arginase 1 Protein (His & MYC Tag) (from HEK293 Cells)
Proteins and Peptides
A DNA sequence encoding the human ARG1 (NP_000036.2) (Met1-Lys322) was expressed with a polyhistidine tag at the N-terminus and a myc tag at the C-terminus.