Human Recombinant TPST1 Protein (His Tag) (from HEK293 Cells)
Supplier: Sino Biological
11258-H07H-50 11258-H07H-50 11258-H07H-20
103680-750EA 191.35 USD
103680-750 103680-752 CA76456-222
Human Recombinant TPST1 Protein (His Tag) (from HEK293 Cells)
Proteins and Peptides
A DNA sequence encoding the human TPST1 (NP_003587.1) extracellular domain (Gln 26-Glu 370) was expressed, with a polyhistidine tag at the N-terminus.