Human Recombinant TrkA/NTRK1 Protein (His & Fc Tag) (from HEK293 Cells)
Supplier: Sino Biological
11073-H03H-50 11073-H03H-100
103680-512EA 191.35 USD
103680-512 103621-892
Human Recombinant TrkA/NTRK1 Protein (His & Fc Tag) (from HEK293 Cells)
Proteins and Peptides
A DNA sequence encoding the human NTRK1 (NP_002520.2) extracellular domain (Met 1-Pro 382) was fused with the C-terminal polyhistidine-tagged Fc region of human IgG1 at the C-terminus.