Human Recombinant respiratory syncytial virus (RSV) (A, rsb1734) glycoprotein G/RSV-G Protein (93% Homology) (His Tag) (from Baculovirus Insect cells)
Supplier: Sino Biological
11070-V08B2-100
103634-404EA 900.2 USD
103634-404
Human Recombinant respiratory syncytial virus (RSV) (A, rsb1734) glycoprotein G/RSV-G Protein (93% Homology) (His Tag) (from Baculovirus Insect cells)
Proteins and Peptides
A DNA sequence encoding the glycoprotein G extracellular domain (Asn 66-Arg297) of human respiratory syncytial virus A (93% homologous with strain rsb1734) (P27022-1) was expressed, with a C-terminal polyhistidine tag.