Influenza B (B/Florida/4/2006) Recombinant Hemagglutinin Protein (HA2 Subunit) (Fc Tag) (from HEK293 Cells)
Supplier: Sino Biological
11053-V04H2-100
103632-678EA 351.48 USD
103632-678
Influenza B (B/Florida/4/2006) Recombinant Hemagglutinin Protein (HA2 Subunit) (Fc Tag) (from HEK293 Cells)
Proteins and Peptides
A DNA sequence encoding the C-terminal fragment (Gly 362-Ala 555) of influenza B virus (B/Florida/4/2006) (ACA33493.1) hemagglutinin, termed as HA2, was fused with the Fc region of mouse IgG1 at the N-terminus.