Human Recombinant KIAA0101/p15/PAF Protein (His Tag) (from E. coli)
Supplier: Sino Biological
10997-H07E-50 10997-H07E-20 10997-H07E-50
103680-342EA 300.98 USD
103680-342 CA76455-874 103680-344
Human Recombinant KIAA0101/p15/PAF Protein (His Tag) (from E. coli)
Proteins and Peptides
A DNA sequence encoding the human PAF isoform 1 (NP_055551.1) (Met 1-Glu 111) was expressed, with a polyhistide tag at the N-terminus.