Human Recombinant EGFL6/EGF-L6 Protein (Flag & His Tag) (from Baculovirus Insect cells)
Supplier: Sino Biological
10867-H18B-100 10867-H18B-20
103680-128EA 191.35 USD
103680-128 103624-124
Human Recombinant EGFL6/EGF-L6 Protein (Flag & His Tag) (from Baculovirus Insect cells)
Proteins and Peptides
A DNA sequence encoding the human EGFL6 (NP_056322.2) (Asn22-Asp553) was expressed with the flag tag at the N-terminus and the histidine tag at the C-terminus.