Human Recombinant AKT2/Protein kinase B/PKB beta Protein (His & GST Tag) (from Baculovirus Insect cells)
Supplier: Sino Biological
10764-H20B-20 10764-H20B-50 10764-H20B-50
103679-918EA 212.85 USD
CA76455-452 103679-918 103679-920
Human Recombinant AKT2/Protein kinase B/PKB beta Protein (His & GST Tag) (from Baculovirus Insect cells)
Proteins and Peptides
A DNA sequence encoding the human AKT2 (NP_001617.1) (Met1-Glu481) was expressed with a C-terminal polyhistidine-tagged GST tag at the N-terminus (his-GST).