Human Recombinant ALK4/ACVR1B Protein (His Tag) (from HEK293 Cells)
Supplier: Sino Biological
10583-H08H-100 10583-H08H-200
103622-520EA 300.98 USD
103622-520 103622-522
Human Recombinant ALK4/ACVR1B Protein (His Tag) (from HEK293 Cells)
Proteins and Peptides
A DNA sequence encoding the human ACVR1B (NP_004293.1) extracellular domain (Met 1-Glu 126) was expressed, with a C-terminal polyhistidine tag.