Human Recombinant IL2RG Protein (His Tag) (from HEK293 Cells)
Supplier: Sino Biological
10555-H08H-100 10555-H08H-1 10555-H08H-1 10555-H08H-100
103906-088EA 351.48 USD
CA76477-570 103906-088 103887-170 CA76479-748
Human Recombinant IL2RG Protein (His Tag) (from HEK293 Cells)
Proteins and Peptides
A DNA sequence encoding the human IL2RG (NP_000197.1) (Met1-Ala262) was expressed with a polyhistide tag at the C-terminus.