Human Recombinant FGFR4/FGF Receptor 4 Protein (Fc Tag) (from HEK293 Cells)
Supplier: Sino Biological
10538-H02H-100 10538-H02H-50
103624-088EA 300.98 USD
103624-088 103678-934
Human Recombinant FGFR4/FGF Receptor 4 Protein (Fc Tag) (from HEK293 Cells)
Proteins and Peptides
A DNA sequence encoding the extracellular domain (Met 1-Asp 369) of human FGFR4 (NP_002002.3) precursor was fused with the Fc region of human IgG1 at the C-terminus.