Human Recombinant MERTK/Mer Protein GST Tag (from Baculovirus Insect cells)
Supplier: Sino Biological
10298-H20B1-20 10298-H20B1-50
103622-400EA 260.58 USD
103622-400 103622-398
Human Recombinant MERTK/Mer Protein GST Tag (from Baculovirus Insect cells)
Proteins and Peptides
A DNA sequence encoding the human MERTK (Q12866) protein kinase domain (Glu 578-Tyr 872) was fused with the N-terminal polyhistidine-tagged GST tag at the N-terminus.